Ricca Chemical Company 2455-16 Cresol Red-Thymol Blue TS (500 mL) (Liquid)

Representative image only.
Ricca Chemical Company Mfr # 2455-16 – Item # EW-86971-68
High quality, competitive pricing, and rapid service
  • Reagents and solutions to meet your lab and research needs
  • CRESOL RED-THYMOL BLUE TS
  • USP
Often bought with…

SPECS & DESCRIPTION

Specifications & Description

  • ClassGeneral Solutions
  • ChemistrySolutions
  • FormLiquid
  • Volume (mL)500
  • PackagingPoly Bottle
  • DOT InformationAlcohols, n.o.s., (Contains: Ethanol and Methanol); Class 3, Packing Group II
  • UN NumberUN1987
  • CAS NumberIncludes 62625-29-0, 64-17-5, 67-56-1, 76-61-9 and 7732-18-5
  • Shelf Life (months)12
  • DescriptionCresol Red-Thymol Blue TS (500 mL) (Liquid)

Key Features

  • Reagents and solutions to meet your lab and research needs
  • CRESOL RED-THYMOL BLUE TS
  • USP

More About this Item

Chemicals sold by Cole-Parmer are not intended for and should not be (i) used for medical, surgical or other patient oriented applications, or (ii) combined with any food product or ingested in any form.


Chemicals sold by Cole-Parmer are not intended for and should not be (i) used for medical, surgical or other patient oriented applications, or (ii) combined with any food product or ingested in any form.

REVIEWS

Q&A

CUSTOMER SUPPORT

Need Help with this product?

Our specialists are here to help you find the best product or part available for your application. Call or Email us and we will make sure you get the right product or part for the job.

Call Us At1-800-323-4340
Live chat