Ricca Chemical Company 2505-16 Cuprous Chloride Reagent, for Cyanide Determination (500 mL)

Representative image only.
Ricca Chemical Company Mfr # 2505-16 – Item # EW-86971-77
  • CUPROUS CHLORIDE REAGENT
Often bought with…

SPECS & DESCRIPTION

Specifications & Description

  • ClassGeneral Solutions
  • ChemistrySolutions
  • FormLiquid
  • Volume (mL)500
  • PackagingAmber Glass
  • DOT InformationCorrosive Liquid, Acidic, Inorganic, n.o.s., (Contains: Hydrochloric Acid); Class 8, Packing Group III
  • CAS NumberIncludes 7440-50-8, 7758-89-6, 7647-01-0 and 7732-18-5
  • UN NumberUN3264
  • Shelf Life (months)12
  • DescriptionCuprous Chloride Reagent, for Cyanide Determination (500 mL)

Key Features

  • CUPROUS CHLORIDE REAGENT

More About this Item


Chemicals sold by Cole-Parmer are not intended for and should not be (i) used for medical, surgical or other patient oriented applications, or (ii) combined with any food product or ingested in any form.

REVIEWS

Q&A

CUSTOMER SUPPORT

Need Help with this product?

Our specialists are here to help you find the best product or part available for your application. Call or Email us and we will make sure you get the right product or part for the job.

Call Us At1-800-323-4340
Live chat