This item is now discontinued. Please browse similar items

Ricca Chemical Company 4622-16 MBTH Indicator, 0.05% (w/v) Aqueous Solution (500 mL)

Representative image only.
Ricca Chemical Company Mfr # 4622-16 – Item # EW-86973-28
  • MBTH INDICATOR, 0.05% (w/v)
  • APHA for Cyanide

SPECS & DESCRIPTION

Specifications & Description

  • Purity GradeComponents all ACS or Reagent
  • ClassGeneral Solutions
  • ChemistrySolutions
  • FormLiquid
  • Volume (mL)500
  • PackagingPoly Bottle
  • DOT InformationNot regulated by DOT
  • CAS NumberIncludes 14448-67-0 and 7732-18-5
  • Shelf Life (months)12
  • DescriptionMBTH Indicator, 0.05% (w/v) Aqueous Solution (500 mL)

Key Features

  • MBTH INDICATOR, 0.05% (w/v)
  • APHA for Cyanide

REVIEWS

Q&A

CUSTOMER SUPPORT

Need Help with this product?

Our specialists are here to help you find the best product or part available for your application. Call or Email us and we will make sure you get the right product or part for the job.

Call Us At1-800-323-4340
Live chat