Ricca Chemical Company 5302-4 Nickel Standard Stock Solution, 1000 ppm Ni (120 mL)

Representative image only.
Ricca Chemical Company Mfr # 5302-4 – Item # EW-86991-44
High quality, competitive pricing, and rapid service
  • Reagents and solutions to meet your lab and research needs
  • NICKEL STOCK STANDARD SOL.
  • EP/ACS 1 mg/mL (1000 ppm Ni)

SPECS & DESCRIPTION

Specifications & Description

  • Concentration1000 ppm
  • Purity GradeComponents all ACS
  • ClassSpectroscopy
  • ChemistryStandards
  • FormLiquid
  • Volume (mL)120
  • PackagingPoly Bottle
  • DOT InformationNot regulated by DOT
  • CAS Number10101-97-0 and 7732-18-5
  • Shelf Life (months)18
  • DescriptionNickel Standard Stock Solution, 1000 ppm Ni (120 mL)

Key Features

  • Reagents and solutions to meet your lab and research needs
  • NICKEL STOCK STANDARD SOL.
  • EP/ACS 1 mg/mL (1000 ppm Ni)

More About this Item

Chemicals sold by Cole-Parmer are not intended for and should not be (i) used for medical, surgical or other patient oriented applications, or (ii) combined with any food product or ingested in any form.


Chemicals sold by Cole-Parmer are not intended for and should not be (i) used for medical, surgical or other patient oriented applications, or (ii) combined with any food product or ingested in any form.

REVIEWS

Q&A

CUSTOMER SUPPORT

Need Help with this product?

Our specialists are here to help you find the best product or part available for your application. Call or Email us and we will make sure you get the right product or part for the job.

Call Us At1-800-323-4340
Live chat