Ricca Chemical Company 5308-32 Nitric Acid, 0.3% (v/v) Aqueous Solution (3 + 997) (1 L)

Representative image only.
Ricca Chemical Company Mfr # 5308-32 – Item # EW-86980-82
High quality, competitive pricing, and rapid service
  • NITRIC ACID, 3 + 997
  • 0.3% v/v Aqueous

PRODUCT OPTIONS

$46.75USD / Each

SPECS & DESCRIPTION

Specifications & Description

  • Concentration0.003
  • Purity GradeComponents all ACS
  • Empirical FormulaHNO3
  • Linear FormulaHNO3
  • ClassGeneral Solutions
  • ChemistryInorganics;Acids
  • FormLiquid
  • Volume (mL)1000
  • Formula Weight63.01
  • PackagingPoly Bottle
  • DOT InformationNot regulated by DOT
  • CAS NumberIncludes 7697-37-2 and 7732-18-5
  • Shelf Life (months)24
  • DescriptionNitric Acid, 0.3% (v/v) Aqueous Solution (3 997) (1 L)

Key Features

  • NITRIC ACID, 3 + 997
  • 0.3% v/v Aqueous

More About this Item

Chemicals sold by Cole-Parmer are not intended for and should not be (i) used for medical, surgical or other patient oriented applications, or (ii) combined with any food product or ingested in any form.


Chemicals sold by Cole-Parmer are not intended for and should not be (i) used for medical, surgical or other patient oriented applications, or (ii) combined with any food product or ingested in any form.

ACCESSORIES

REVIEWS

Q&A

CUSTOMER SUPPORT

Need Help with this product?

Our specialists are here to help you find the best product or part available for your application. Call or Email us and we will make sure you get the right product or part for the job.

Call Us At1-800-323-4340
Live chat