Ricca Chemical Company 5464.5-16 ORP Standard, +200 mV vs. Ag/AgCl (4M KCl sat. with AgCl filling solution) (500 mL)

Representative image only.
Ricca Chemical Company Mfr # 5464.5-16 – Item # EW-86974-09
Fast, simple calibration
  • Use as relative mV standard and calibrate offset of sample

PRODUCT OPTIONS

$58.00USD / Each

SPECS & DESCRIPTION

Specifications & Description

  • Purity GradeComponents all ACS
  • ClassStandards
  • ChemistryStandards
  • FormLiquid
  • Volume (mL)500
  • PackagingAmber Glass
  • DOT InformationNot regulated by DOT
  • CAS NumberIncludes 7447-40-7, 13746-66-2, 14459-95-1 and 7732-18-5
  • Shelf Life (months)9
  • DescriptionORP Standard, +200 mV vs. Ag/AgCl (4M KCl sat. with AgCl filling solution) (500 mL)

Key Features

  • Use as relative mV standard and calibrate offset of sample

More About this Item

This ORP standard features +200 mV vs. Ag/AgCl with 4M KCl saturated with AgCl filling solution.

Chemicals sold by Cole-Parmer are not intended for and should not be (i) used for medical, surgical or other patient oriented applications, or (ii) combined with any food product or ingested in any form.

REVIEWS

Q&A

CUSTOMER SUPPORT

Need Help with this product?

Our specialists are here to help you find the best product or part available for your application. Call or Email us and we will make sure you get the right product or part for the job.

Call Us At1-800-323-4340
Live chat