Ricca Chemical Company 6220-5 Potassium Hydroxide, 0.250 Normal (N/4) (20 L)

Representative image only.
Ricca Chemical Company Mfr # 6220-5 – Item # EW-86993-81
  • POT. HYDROXIDE, 0.250 Normal

PRODUCT OPTIONS

$135.00USD / Each
Often bought with…

SPECS & DESCRIPTION

Specifications & Description

  • Normality0.25
  • Purity GradeComponents all ACS
  • ClassTitrants
  • ChemistrySolutions
  • FormLiquid
  • Volume (mL)20000
  • PackagingCubitainer
  • DOT InformationNot regulated by DOT
  • CAS NumberIncludes 1310-58-3 and 7732-18-5
  • Shelf Life (months)24
  • DescriptionPotassium Hydroxide, 0.250 Normal (N/4) (20 L)

Key Features

  • POT. HYDROXIDE, 0.250 Normal

More About this Item


Chemicals sold by Cole-Parmer are not intended for and should not be (i) used for medical, surgical or other patient oriented applications, or (ii) combined with any food product or ingested in any form.

ACCESSORIES

REVIEWS

Q&A

CUSTOMER SUPPORT

Need Help with this product?

Our specialists are here to help you find the best product or part available for your application. Call or Email us and we will make sure you get the right product or part for the job.

Call Us At1-800-323-4340
Live chat