Ricca Chemical Company 6260-1 Potassium Hydroxide, 0.100 Normal (N/10) in Methanol (1 gal)

Representative image only.
Ricca Chemical Company Mfr # 6260-1 – Item # EW-86986-96
Quality and consistency your lab can count on
  • Reagents and solutions to meet your lab and research needs
  • POT. HYDROXIDE, 0.100 N/MEOH
Often bought with…

SPECS & DESCRIPTION

Specifications & Description

  • Normality0.1
  • Purity GradeComponents all ACS
  • ClassTitrants
  • ChemistrySolvents
  • FormLiquid
  • Volume (mL)3785
  • PackagingPoly Bottle
  • DOT InformationFlammable Liquid, n.o.s., (Contains: Methanol); Class 3, Packing Group II
  • CAS NumberIncludes 67-56-1 and 1310-58-3
  • UN NumberUN1993
  • Shelf Life (months)12
  • DescriptionPotassium Hydroxide, 0.100 Normal (N/10) in Methanol (1 gal)

Key Features

  • Reagents and solutions to meet your lab and research needs
  • POT. HYDROXIDE, 0.100 N/MEOH

More About this Item

Chemicals sold by Cole-Parmer are not intended for and should not be (i) used for medical, surgical or other patient oriented applications, or (ii) combined with any food product or ingested in any form.


Chemicals sold by Cole-Parmer are not intended for and should not be (i) used for medical, surgical or other patient oriented applications, or (ii) combined with any food product or ingested in any form.

REVIEWS

Q&A

CUSTOMER SUPPORT

Need Help with this product?

Our specialists are here to help you find the best product or part available for your application. Call or Email us and we will make sure you get the right product or part for the job.

Call Us At1-800-323-4340
Live chat