Ricca Chemical Company 6266.9-1 Potassium Hydroxide, 1.00 Normal in Denatured Alcohol (4 L)

Representative image only.
Ricca Chemical Company Mfr # 6266.9-1 – Item # EW-86987-01
  • POT. HYDROXIDE, 1.00 N/SDA
Often bought with…

SPECS & DESCRIPTION

Specifications & Description

  • Normality1
  • ClassTitrants
  • ChemistrySolutions
  • FormLiquid
  • Volume (mL)4000
  • PackagingPoly Bottle
  • DOT InformationFlammable Liquid, Corrosive, n.o.s., (Contains: Ethanol and Potassium Hydroxide); Class 8 (3), Packing Group II
  • UN NumberUN2924
  • CAS NumberIncludes 64-17-5, 67-56-1, 1310-58-3 and proprietary preservative.
  • Shelf Life (months)12
  • DescriptionPotassium Hydroxide, 1.00 Normal in Denatured Alcohol (4 L)

Key Features

  • POT. HYDROXIDE, 1.00 N/SDA

More About this Item


Chemicals sold by Cole-Parmer are not intended for and should not be (i) used for medical, surgical or other patient oriented applications, or (ii) combined with any food product or ingested in any form.

REVIEWS

Q&A

CUSTOMER SUPPORT

Need Help with this product?

Our specialists are here to help you find the best product or part available for your application. Call or Email us and we will make sure you get the right product or part for the job.

Call Us At1-800-323-4340
Live chat