Ricca Chemical Company 6630-16 Robert's Reagent, for the Qualitative Detection of Albumin in Urine (500 mL)

Representative image only.
Ricca Chemical Company Mfr # 6630-16 – Item # EW-86975-36
  • ROBERT'S REAGENT

SPECS & DESCRIPTION

Specifications & Description

  • Purity GradeComponents all ACS
  • ClassGeneral Solutions
  • ChemistrySolutions
  • FormLiquid
  • Volume (mL)500
  • PackagingAmber Glass
  • DOT InformationNitric Acid Solution; Class 8, Packing Group II
  • UN NumberUN2031
  • CAS NumberIncludes 10034-99-8, 7697-37-2 and 7732-18-5
  • Shelf Life (months)24
  • DescriptionRobert's Reagent, for the Qualitative Detection of Albumin in Urine (500 mL)

Key Features

  • ROBERT'S REAGENT

More About this Item


Chemicals sold by Cole-Parmer are not intended for and should not be (i) used for medical, surgical or other patient oriented applications, or (ii) combined with any food product or ingested in any form.

REVIEWS

Q&A

CUSTOMER SUPPORT

Need Help with this product?

Our specialists are here to help you find the best product or part available for your application. Call or Email us and we will make sure you get the right product or part for the job.

Call Us At1-800-323-4340
Live chat