Ricca Chemical Company 6655-5 Saccomanno Collection Fluid, Fixative and Preservative for Sputum Samples (20 L)

Representative image only.
Ricca Chemical Company Mfr # 6655-5 – Item # EW-86993-94
  • SACCOMANNO COLLECTION FLUID

SPECS & DESCRIPTION

Specifications & Description

  • ClassGeneral Solutions
  • ChemistrySolutions
  • FormLiquid
  • Volume (mL)20000
  • PackagingCubitainer
  • DOT InformationAlcohols, N.O.S.., (Contains: Ethanol and Methanol); Class 3, Packing Group III
  • UN NumberUN1987
  • CAS NumberIncludes 64-17-5, 67-56-1, 25322-68-3, a proprietary preservative, and 7732-18-5
  • Shelf Life (months)18
  • DescriptionSaccomanno Collection Fluid, Fixative and Preservative for Sputum Samples (20 L)

Key Features

  • SACCOMANNO COLLECTION FLUID

REVIEWS

Q&A

CUSTOMER SUPPORT

Need Help with this product?

Our specialists are here to help you find the best product or part available for your application. Call or Email us and we will make sure you get the right product or part for the job.

Call Us At1-800-323-4340
Live chat