Ricca Chemical Company 6680-16 Safranine, Aqueous-Alcoholic, Counterstain for Gram Staining (500 mL)

Representative image only.
Ricca Chemical Company Mfr # 6680-16 – Item # EW-86975-41
  • SAFRANINE, Aqueous-Alcoholic

PRODUCT OPTIONS

$35.50USD / Each

SPECS & DESCRIPTION

Specifications & Description

  • ClassGeneral Solutions
  • ChemistrySolutions
  • FormLiquid
  • Volume (mL)500
  • PackagingPoly Bottle
  • DOT InformationNot regulated by DOT
  • CAS NumberIncludes 64-17-5, 67-56-1, 477-73-6 and 7732-18-5
  • Shelf Life (months)24
  • DescriptionSafranine, Aqueous-Alcoholic, Counterstain for Gram Staining (500 mL)

Key Features

  • SAFRANINE, Aqueous-Alcoholic

ACCESSORIES

REVIEWS

Q&A

CUSTOMER SUPPORT

Need Help with this product?

Our specialists are here to help you find the best product or part available for your application. Call or Email us and we will make sure you get the right product or part for the job.

Call Us At1-800-323-4340
Live chat