Ricca Chemical Company 6710-16 Semen Diluting Fluid, Bicarbonate-Formalin Diluting Fluid for Sperm Count (500 mL)

Representative image only.
Ricca Chemical Company Mfr # 6710-16 – Item # EW-86975-43
  • SEMEN DILUTING FLUID

SPECS & DESCRIPTION

Specifications & Description

  • Purity GradeComponents all ACS
  • ClassGeneral Solutions
  • ChemistrySolutions
  • FormLiquid
  • Volume (mL)500
  • PackagingPoly Bottle
  • DOT InformationNot regulated by DOT
  • CAS NumberIncludes 50-00-0, 144-55-8 and 7732-18-5
  • Shelf Life (months)18
  • DescriptionSemen Diluting Fluid, Bicarbonate-Formalin Diluting Fluid for Sperm Count (500 mL)

Key Features

  • SEMEN DILUTING FLUID

REVIEWS

Q&A

CUSTOMER SUPPORT

Need Help with this product?

Our specialists are here to help you find the best product or part available for your application. Call or Email us and we will make sure you get the right product or part for the job.

Call Us At1-800-323-4340
Live chat