Ricca Chemical Company 6810-4 Silver Diethyldithiocarbamate, 0.3% (w/v) in Chloroform (120 mL)

Representative image only.
Ricca Chemical Company Mfr # 6810-4 – Item # EW-86992-09
  • SILV DIETHYLDITHIOCARBAMATE
  • 0.3% in Chloroform/APHA-As

SPECS & DESCRIPTION

Specifications & Description

  • Purity GradeComponents all ACS
  • ClassGeneral Solutions
  • ChemistrySolutions
  • FormLiquid
  • Volume (mL)120
  • PackagingAmber Glass
  • DOT InformationChloroform Solution; Class 6.1, Packing Group III
  • CAS NumberIncludes 67-66-3, 110-91-8 and 1470-61-7
  • UN NumberUN1888
  • Shelf Life (months)12
  • DescriptionSilver Diethyldithiocarbamate, 0.3% (w/v) in Chloroform (120 mL)

Key Features

  • SILV DIETHYLDITHIOCARBAMATE
  • 0.3% in Chloroform/APHA-As

More About this Item


Chemicals sold by Cole-Parmer are not intended for and should not be (i) used for medical, surgical or other patient oriented applications, or (ii) combined with any food product or ingested in any form.

REVIEWS

Q&A

CUSTOMER SUPPORT

Need Help with this product?

Our specialists are here to help you find the best product or part available for your application. Call or Email us and we will make sure you get the right product or part for the job.

Call Us At1-800-323-4340
Live chat