Ricca Chemical Company 6850-2.5 Silver Nitrate, 0.0100 Normal (N/100) (10 L)

Representative image only.
Ricca Chemical Company Mfr # 6850-2.5 – Item # EW-86989-03
High quality, competitive pricing, and rapid service
  • Reagents and solutions to meet your lab and research needs
  • SILVER NITRATE, 0.0100 N

SPECS & DESCRIPTION

Specifications & Description

  • Normality0.01
  • Purity GradeComponents all ACS
  • Linear FormulaAgNO3
  • ClassTitrants
  • ChemistrySolutions
  • FormLiquid
  • SynonymsNitric acid, silver salt
  • Volume (mL)10000
  • Formula Weight169.87
  • PackagingCubitainer
  • DOT InformationNot regulated by DOT
  • CAS NumberIncludes 7761-88-8 and 7732-18-5
  • Shelf Life (months)18
  • DescriptionSilver Nitrate, 0.0100 Normal (N/100) (10 L)

Key Features

  • Reagents and solutions to meet your lab and research needs
  • SILVER NITRATE, 0.0100 N

More About this Item

Chemicals sold by Cole-Parmer are not intended for and should not be (i) used for medical, surgical or other patient oriented applications, or (ii) combined with any food product or ingested in any form.


Chemicals sold by Cole-Parmer are not intended for and should not be (i) used for medical, surgical or other patient oriented applications, or (ii) combined with any food product or ingested in any form.

REVIEWS

Q&A

CUSTOMER SUPPORT

Need Help with this product?

Our specialists are here to help you find the best product or part available for your application. Call or Email us and we will make sure you get the right product or part for the job.

Call Us At1-800-323-4340
Live chat