Ricca Chemical Company 7027-32 Silver Nitrate, 0.0100 Normal (N/100) in Denatured Alcohol (1 L)

Representative image only.
Ricca Chemical Company Mfr # 7027-32 – Item # EW-86982-42
High quality, competitive pricing, and rapid service
  • Reagents and solutions to meet your lab and research needs
  • SILVER NITRATE, 0.01 N/SDA

SPECS & DESCRIPTION

Specifications & Description

  • Normality0.01
  • Linear FormulaAgNO3
  • ClassTitrants
  • ChemistrySolutions
  • FormLiquid
  • SynonymsNitric acid, silver salt
  • Volume (mL)1000
  • Formula Weight169.87
  • PackagingAmber Poly Bottle
  • DOT InformationAlcohols, N.O.S., (Ethanol and Methanol); Class 3, Packing Group II
  • UN NumberUN1987
  • CAS NumberIncludes 64-17-5, 67-56-1, and 7761-88-8
  • Shelf Life (months)12
  • DescriptionSilver Nitrate, 0.0100 Normal (N/100) in Denatured Alcohol (1 L)

Key Features

  • Reagents and solutions to meet your lab and research needs
  • SILVER NITRATE, 0.01 N/SDA

More About this Item

Chemicals sold by Cole-Parmer are not intended for and should not be (i) used for medical, surgical or other patient oriented applications, or (ii) combined with any food product or ingested in any form.


Chemicals sold by Cole-Parmer are not intended for and should not be (i) used for medical, surgical or other patient oriented applications, or (ii) combined with any food product or ingested in any form.

REVIEWS

Q&A

CUSTOMER SUPPORT

Need Help with this product?

Our specialists are here to help you find the best product or part available for your application. Call or Email us and we will make sure you get the right product or part for the job.

Call Us At1-800-323-4340
Live chat