Ricca Chemical Company 7194-16 Sodium Chloride-Hydroxylamine Sulfate f/ Cold Vapor Atomic Absorption, 500 mL

Representative image only.
Ricca Chemical Company Mfr # 7194-16 – Item # EW-86975-79
  • NaCl-HYDROXYLAMINE SULFATE
  • APHA for Metals/ASTM D 3223

SPECS & DESCRIPTION

Specifications & Description

  • Purity GradeComponents all ACS
  • ClassGeneral Solutions
  • ChemistrySolutions
  • FormLiquid
  • Volume (mL)500
  • PackagingPoly Bottle
  • DOT InformationNot regulated by DOT
  • CAS NumberIncludes 10039-54-0, 7647-14-5, and 7732-18-5
  • Shelf Life (months)12
  • DescriptionSodium Chloride-Hydroxylamine Sulfate f/ Cold Vapor Atomic Absorption, 500 mL

Key Features

  • NaCl-HYDROXYLAMINE SULFATE
  • APHA for Metals/ASTM D 3223

More About this Item

Sodium Chloride-Hydroxylamine Sulfate Solution, for Cold Vapor Atomic Absorption Analysis


Chemicals sold by Cole-Parmer are not intended for and should not be (i) used for medical, surgical or other patient oriented applications, or (ii) combined with any food product or ingested in any form.

REVIEWS

Q&A

CUSTOMER SUPPORT

Need Help with this product?

Our specialists are here to help you find the best product or part available for your application. Call or Email us and we will make sure you get the right product or part for the job.

Call Us At1-800-323-4340
Live chat