Ricca Chemical Company 7212-32 Sodium Chloride, 10 g/L in Alcohol, Salts in Crude Oil; 32 oz

Representative image only.
Ricca Chemical Company Mfr # 7212-32 – Item # EW-86982-67
  • SODIUM CHLORIDE, 10 gpL in
  • 75% (v/v) Mixed Alcohol Solv
  • For determination of Salts in Crude Oil

SPECS & DESCRIPTION

Specifications & Description

  • Purity GradeComponents all ACS
  • ClassGeneral Solutions
  • ChemistrySolutions
  • FormLiquid
  • Volume (mL)1000
  • PackagingAmber Glass
  • DOT InformationAlcohols, N.O.S., (Butanol and Methanol); Class 3, Packing Group II
  • UN NumberUN1987
  • CAS NumberIncludes 64-56-1, 71-36-3, 7647-14-5, and 7732-18-5
  • Shelf Life (months)12
  • DescriptionSodium Chloride, 10 g/L in Alcohol, Salts in Crude Oil; 32 oz

Key Features

  • SODIUM CHLORIDE, 10 gpL in
  • 75% (v/v) Mixed Alcohol Solv
  • For determination of Salts in Crude Oil

More About this Item

Sodium Chloride, 10 g/L in Alcohol, Salts in Crude Oil; 32 oz


Chemicals sold by Cole-Parmer are not intended for and should not be (i) used for medical, surgical or other patient oriented applications, or (ii) combined with any food product or ingested in any form.

REVIEWS

Q&A

CUSTOMER SUPPORT

Need Help with this product?

Our specialists are here to help you find the best product or part available for your application. Call or Email us and we will make sure you get the right product or part for the job.

Call Us At1-800-323-4340
Live chat