Ricca Chemical Company 7377-5 Sodium Hydroxide, 0.333 Normal (N/3) (20 L)

Representative image only.
Ricca Chemical Company Mfr # 7377-5 – Item # EW-86994-25
High quality, competitive pricing, and rapid service
  • Reagents and solutions to meet your lab and research needs
  • SODIUM HYDROXIDE, 0.333 N

SPECS & DESCRIPTION

Specifications & Description

  • Normality0.333
  • Purity GradeComponents all ACS
  • ClassTitrants
  • ChemistrySolutions
  • FormLiquid
  • SynonymsCaustic Soda Solution
  • Volume (mL)20000
  • PackagingCubitainer
  • DOT InformationSodium Hydroxide Solution; Class 8, Packing Group III
  • CAS NumberIncludes 1310-73-2 and 7732-18-5
  • UN NumberUN1824
  • Shelf Life (months)24
  • DescriptionSodium Hydroxide, 0.333 Normal (N/3) (20 L)

Key Features

  • Reagents and solutions to meet your lab and research needs
  • SODIUM HYDROXIDE, 0.333 N

More About this Item

Chemicals sold by Cole-Parmer are not intended for and should not be (i) used for medical, surgical or other patient oriented applications, or (ii) combined with any food product or ingested in any form.


Chemicals sold by Cole-Parmer are not intended for and should not be (i) used for medical, surgical or other patient oriented applications, or (ii) combined with any food product or ingested in any form.

REVIEWS

Q&A

CUSTOMER SUPPORT

Need Help with this product?

Our specialists are here to help you find the best product or part available for your application. Call or Email us and we will make sure you get the right product or part for the job.

Call Us At1-800-323-4340
Live chat