Ricca Chemical Company 7459-32 Sodium Hydroxide, 3.00 Normal (1 L) (Liquid)

Representative image only.
Ricca Chemical Company Mfr # 7459-32 – Item # EW-86983-01
Quality and consistency your lab can count on
  • SODIUM HYDROXIDE, 3.00 N

SPECS & DESCRIPTION

Specifications & Description

  • Normality3
  • Purity GradeComponents all ACS
  • ClassGeneral Solutions
  • ChemistrySolutions
  • FormLiquid
  • SynonymsCaustic Soda Solution
  • Volume (mL)1000
  • PackagingPoly Bottle
  • DOT InformationSodium Hydroxide Solution; Class 8, Packing Group II
  • CAS NumberIncludes 1310-73-2 and 7732-18-5
  • UN NumberUN1824
  • Shelf Life (months)24
  • DescriptionSodium Hydroxide, 3.00 Normal (1 L) (Liquid)

Key Features

  • SODIUM HYDROXIDE, 3.00 N

More About this Item

Chemicals sold by Cole-Parmer are not intended for and should not be (i) used for medical, surgical or other patient oriented applications, or (ii) combined with any food product or ingested in any form.


Chemicals sold by Cole-Parmer are not intended for and should not be (i) used for medical, surgical or other patient oriented applications, or (ii) combined with any food product or ingested in any form.

REVIEWS

Q&A

CUSTOMER SUPPORT

Need Help with this product?

Our specialists are here to help you find the best product or part available for your application. Call or Email us and we will make sure you get the right product or part for the job.

Call Us At1-800-323-4340
Live chat