Ricca Chemical Company 7522-16 Sodium Phosphate, Dibasic TS, 12% (w/v) Aqueous Solution (500 mL)

Representative image only.
Ricca Chemical Company Mfr # 7522-16 – Item # EW-86976-18
Quality and consistency your lab can count on
  • Reagents and solutions to meet your lab and research needs
  • SODIUM PHOSPHATE, DIBASIC TS
  • 12% (w/v)

SPECS & DESCRIPTION

Specifications & Description

  • Purity GradeComponents all ACS
  • ClassGeneral Solutions
  • ChemistrySolutions
  • FormLiquid
  • Volume (mL)500
  • PackagingPoly Bottle
  • DOT InformationNot regulated by DOT
  • CAS NumberIncludes 7558-79-4 and 7732-18-5
  • Shelf Life (months)18
  • DescriptionSodium Phosphate, Dibasic TS, 12% (w/v) Aqueous Solution (500 mL)

Key Features

  • Reagents and solutions to meet your lab and research needs
  • SODIUM PHOSPHATE, DIBASIC TS
  • 12% (w/v)

More About this Item

Chemicals sold by Cole-Parmer are not intended for and should not be (i) used for medical, surgical or other patient oriented applications, or (ii) combined with any food product or ingested in any form.


Chemicals sold by Cole-Parmer are not intended for and should not be (i) used for medical, surgical or other patient oriented applications, or (ii) combined with any food product or ingested in any form.

REVIEWS

Q&A

CUSTOMER SUPPORT

Need Help with this product?

Our specialists are here to help you find the best product or part available for your application. Call or Email us and we will make sure you get the right product or part for the job.

Call Us At1-800-323-4340
Live chat