Ricca Chemical Company 7552-4 Sodium Potassium Tartrate, 30% (w/v) Aqueous Solution (120 mL)

Representative image only.
Ricca Chemical Company Mfr # 7552-4 – Item # EW-86992-23
  • SOD. POT. TARTRATE, 30%

SPECS & DESCRIPTION

Specifications & Description

  • Purity GradeComponents all ACS
  • ClassGeneral Solutions
  • ChemistrySolutions
  • FormLiquid
  • Volume (mL)120
  • PackagingPoly Bottle
  • DOT InformationNot regulated by DOT
  • CAS NumberIncludes 6381-59-5 and 7732-18-5
  • Shelf Life (months)24
  • DescriptionSodium Potassium Tartrate, 30% (w/v) Aqueous Solution (120 mL)

Key Features

  • SOD. POT. TARTRATE, 30%

More About this Item


Chemicals sold by Cole-Parmer are not intended for and should not be (i) used for medical, surgical or other patient oriented applications, or (ii) combined with any food product or ingested in any form.

REVIEWS

Q&A

CUSTOMER SUPPORT

Need Help with this product?

Our specialists are here to help you find the best product or part available for your application. Call or Email us and we will make sure you get the right product or part for the job.

Call Us At1-800-323-4340
Live chat