Ricca Chemical Company 8180-16 Sulfuric Acid, 50% (v/v) Aqueous Solution (1 + 1) (500 mL)

Representative image only.
Ricca Chemical Company Mfr # 8180-16 – Item # EW-86976-69
Quality and consistency your lab can count on
  • Reagents and solutions to meet your lab and research needs
  • SULFURIC ACID, 50% (v/v)

PRODUCT OPTIONS

$54.00USD / Each

SPECS & DESCRIPTION

Specifications & Description

  • ClassGeneral Solutions
  • ChemistryInorganics;Acids
  • FormLiquid
  • SynonymsHygrogen sulfate; Oil of vitriol; Vitriol brown oil; Mattling acid; Battery acid; Sulphuric acid; Electrolyte acid; Dihydrogen sulfate; Spirt of sulphur; Chamber acid
  • Volume (mL)500
  • PackagingPoly Bottle
  • DOT InformationSulphuric Acid Solution; Class 8, Packing Group II
  • UN NumberUN1830
  • CAS NumberIncludes 7664-93-9 and 7732-18-5
  • Shelf Life (months)24
  • DescriptionSulfuric Acid, 50% (v/v) Aqueous Solution (1 + 1) (500 mL)

Key Features

  • Reagents and solutions to meet your lab and research needs
  • SULFURIC ACID, 50% (v/v)

More About this Item

Chemicals sold by Cole-Parmer are not intended for and should not be (i) used for medical, surgical or other patient oriented applications, or (ii) combined with any food product or ingested in any form.


Chemicals sold by Cole-Parmer are not intended for and should not be (i) used for medical, surgical or other patient oriented applications, or (ii) combined with any food product or ingested in any form.

REVIEWS

Q&A

CUSTOMER SUPPORT

Need Help with this product?

Our specialists are here to help you find the best product or part available for your application. Call or Email us and we will make sure you get the right product or part for the job.

Call Us At1-800-323-4340
Live chat