Ricca Chemical Company 9150-32 Water, ACS Reagent Grade, ASTM Type I (1 L)

Representative image only.
Ricca Chemical Company Mfr # 9150-32 – Item # EW-86983-98
  • WATER, Reagent Grade

PRODUCT OPTIONS

$20.50USD / Each

SPECS & DESCRIPTION

Specifications & Description

  • Purity GradeACS Reagent
  • ClassHigh Purity Solutions (Water)
  • FormLiquid
  • SynonymsType I Water, DI Water
  • Volume (mL)1000
  • PackagingPoly Bottle
  • DOT InformationNot regulated by DOT
  • CAS Number7732-18-5
  • Shelf Life (months)12
  • DescriptionWater, ACS Reagent Grade, ASTM Type I (1 L)

Key Features

  • WATER, Reagent Grade

More About this Item


Chemicals sold by Cole-Parmer are not intended for and should not be (i) used for medical, surgical or other patient oriented applications, or (ii) combined with any food product or ingested in any form.

ACCESSORIES

REVIEWS

Q&A

CUSTOMER SUPPORT

Need Help with this product?

Our specialists are here to help you find the best product or part available for your application. Call or Email us and we will make sure you get the right product or part for the job.

Call Us At1-800-323-4340
Live chat