Ricca Chemical Company 9201-32 Water Hardness Buffer, without Magnesium EDTA (1 L)

Representative image only.
Ricca Chemical Company Mfr # 9201-32 – Item # EW-86984-02
  • WATER HARDNESS BUFFER
  • w/o Magnesium

$54.50USD / Each

SPECS & DESCRIPTION

Specifications & Description

  • Purity GradeComponents all ACS
  • ClassBuffers
  • FormLiquid
  • Volume (mL)1000
  • PackagingPoly Bottle
  • DOT InformationAmmonia solution; Class 8, Packing Group III
  • CAS NumberIncludes 12125-02-9, 1336-21-6, and 7732-15-5
  • UN NumberUN2672
  • Shelf Life (months)12
  • DescriptionWater Hardness Buffer, without Magnesium EDTA (1 L)

Key Features

  • WATER HARDNESS BUFFER
  • w/o Magnesium

More About this Item


Chemicals sold by Cole-Parmer are not intended for and should not be (i) used for medical, surgical or other patient oriented applications, or (ii) combined with any food product or ingested in any form.

REVIEWS

Q&A

CUSTOMER SUPPORT

Need Help with this product?

Our specialists are here to help you find the best product or part available for your application. Call or Email us and we will make sure you get the right product or part for the job.

Call Us At1-800-323-4340
Live chat