Ricca Chemical Company 9301-4 Water Hardness Indicator, 1% (w/v) Eriochrome Black T in 20% (v/v) (120 mL)

Representative image only.
Ricca Chemical Company Mfr # 9301-4 – Item # EW-86992-77
Quality and consistency your lab can count on
  • Reagents and solutions to meet your lab and research needs
  • WATER HARDNESS INDICATOR
  • APHA, Modified

$42.00USD / Each

SPECS & DESCRIPTION

Specifications & Description

  • Purity GradeComponents all ACS or Reagent
  • ClassGeneral Solutions
  • FormLiquid
  • Volume (mL)120
  • PackagingAmber Glass
  • DOT InformationFlammable Liquid, Corrosive, n.o.s., (Isopropanol and Triethanolamine); Class 3 (8), Packing Group II
  • CAS NumberIncludes 1787-61-7, 67-63-0, and 102-71-6
  • UN NumberUN2924
  • Shelf Life (months)12
  • DescriptionWater Hardness Indicator, 1% (w/v) Eriochrome Black T in 20% (v/v) (120 mL)

Key Features

  • Reagents and solutions to meet your lab and research needs
  • WATER HARDNESS INDICATOR
  • APHA, Modified

More About this Item

Chemicals sold by Cole-Parmer are not intended for and should not be (i) used for medical, surgical or other patient oriented applications, or (ii) combined with any food product or ingested in any form.


Chemicals sold by Cole-Parmer are not intended for and should not be (i) used for medical, surgical or other patient oriented applications, or (ii) combined with any food product or ingested in any form.

REVIEWS

Q&A

CUSTOMER SUPPORT

Need Help with this product?

Our specialists are here to help you find the best product or part available for your application. Call or Email us and we will make sure you get the right product or part for the job.

Call Us At1-800-323-4340
Live chat