Ricca Chemical Company ACU1KN-500 Copper Standard, 1 mL = 1 mg Cu, 1000 ppm Cu (500 mL)

Representative image only.
Ricca Chemical Company Mfr # ACU1KN-500 – Item # EW-86971-58
Quality and consistency your lab can count on
  • Reagents and solutions to meet your lab and research needs
  • COPPER STD., 1 = 1 Cu
  • Equivalent to 1,000 ppm
Often bought with…

SPECS & DESCRIPTION

Specifications & Description

  • ClassSpectroscopy
  • ChemistryStandards
  • FormLiquid
  • Volume (mL)500
  • PackagingPoly Bottle
  • DOT InformationCorrosive Liquid, Acidic, Inorganic, n.o.s., (Nitric Acid); Class 8, Packing Group III
  • UN NumberUN3264
  • CAS NumberIncludes 7440-50-8, 7697-37-2 and 7732-18-5
  • Shelf Life (months)24
  • DescriptionCopper Standard, 1 mL = 1 mg Cu, 1000 ppm Cu (500 mL)

Key Features

  • Reagents and solutions to meet your lab and research needs
  • COPPER STD., 1 = 1 Cu
  • Equivalent to 1,000 ppm

More About this Item

Chemicals sold by Cole-Parmer are not intended for and should not be (i) used for medical, surgical or other patient oriented applications, or (ii) combined with any food product or ingested in any form.


Chemicals sold by Cole-Parmer are not intended for and should not be (i) used for medical, surgical or other patient oriented applications, or (ii) combined with any food product or ingested in any form.

REVIEWS

Q&A

CUSTOMER SUPPORT

Need Help with this product?

Our specialists are here to help you find the best product or part available for your application. Call or Email us and we will make sure you get the right product or part for the job.

Call Us At1-800-323-4340
Live chat