Ricca Chemical Company AMN1KN-500 Manganese Standard, 1 mL = 1 mg Mn, 1000 ppm Mn (500 mL)

Representative image only.
Ricca Chemical Company Mfr # AMN1KN-500 – Item # EW-86973-24
Quality and consistency your lab can count on
  • 1 ml = 0.05 mg Mn
  • MANGANESE STD., 1 = 1 Mn
  • Equivalent to 1,000 ppm

SPECS & DESCRIPTION

Specifications & Description

  • ClassSpectroscopy
  • ChemistryStandards
  • FormLiquid
  • Volume (mL)500
  • PackagingPoly Bottle
  • DOT InformationCorrosive Liquid, Acidic, Inorganic, n.o.s., (Nitric Acid); Class 8, Packing Group III
  • CAS NumberIncludes 6156-78-1, 7697-37-2 and 7732-18-6
  • UN NumberUN3264
  • Shelf Life (months)24
  • DescriptionManganese Standard, 1 mL = 1 mg Mn, 1000 ppm Mn (500 mL)

Key Features

  • 1 ml = 0.05 mg Mn
  • MANGANESE STD., 1 = 1 Mn
  • Equivalent to 1,000 ppm

More About this Item

Chemicals sold by Cole-Parmer are not intended for and should not be (i) used for medical, surgical or other patient oriented applications, or (ii) combined with any food product or ingested in any form.


Chemicals sold by Cole-Parmer are not intended for and should not be (i) used for medical, surgical or other patient oriented applications, or (ii) combined with any food product or ingested in any form.

REVIEWS

Q&A

CUSTOMER SUPPORT

Need Help with this product?

Our specialists are here to help you find the best product or part available for your application. Call or Email us and we will make sure you get the right product or part for the job.

Call Us At1-800-323-4340
Live chat