Ricca Chemical Company Distilled Water, Reagent Grade, 1 L

Representative image only.
Ricca Chemical Company Mfr # 9180-32 – Item # UX-86984-00
Quality and consistency your lab can count on
  • Water meets ACS Reagent grade standards
  • Water comes in 1 L size and packaged in an amber glass bottle

PRODUCT OPTIONS

$54.70USD / Each

SPECS & DESCRIPTION

Specifications & Description

  • Purity GradeReagent
  • Empirical FormulaH2O
  • FormLiquid
  • Synonymsdihydrogen oxide, dihydrogen monoxide
  • Volume (mL)1000
  • Formula Weight18.02
  • PackagingAmber Glass Bottle
  • DOT InformationNot regulated by DOT
  • CAS Number7723-18-5
  • Shelf Life (months)18
  • Boiling Point C (° C)100
  • Boiling Point F (° F)212
  • DescriptionDistilled Water, Reagent Grade, 1 L

Key Features

  • Water meets ACS Reagent grade standards
  • Water comes in 1 L size and packaged in an amber glass bottle

More About this Item

Chemicals sold by Cole-Parmer are not intended for and should not be (i) used for medical, surgical or other patient oriented applications, or (ii) combined with any food product or ingested in any form.

REVIEWS

Q&A

CUSTOMER SUPPORT

Need Help with this product?

Our specialists are here to help you find the best product or part available for your application. Call or Email us and we will make sure you get the right product or part for the job.

Call Us At1-800-323-4340
Live chat