Ricca Chemical Company HydroSpec™ Coulometric CG Catholyte Solution, cells with a diaphragm; 6 x 110 mL

Representative image only.
Ricca Chemical Company Mfr # RK130000-6x110C – Item # EW-86994-89
Specially formulated reagents produce high performance and reliable results
  • Support a wide range of sample types from ethanol and fuels to fats and oils
  • Available for titration for cells with or without a diaphragm
Often bought with…

SPECS & DESCRIPTION

Specifications & Description

  • ClassReagent Solvents
  • FormLiquid
  • Volume (mL)660
  • PackagingAmber Glass
  • DOT InformationMethanol Solution; Class 3, Packing Group II
  • UN NumberUN1230
  • CAS NumberIncludes 111-90-0, 67-56-1, 288-32-4, 7446-09-5, 68007-08-9
  • Shelf Life (days)1800
  • DescriptionCoulometric CG Catholyte Solution, cells with a diaphragm
CorrosiveFlammableSolventToxic

Key Features

  • Support a wide range of sample types from ethanol and fuels to fats and oils
  • Available for titration for cells with or without a diaphragm

More About this Item

These highly stable solvents retain activity for up to 5 years. 1 L bottles have a GL45 threaded neck to make connection to Karl Fischer titrators easy and simple.

REVIEWS

Q&A

CUSTOMER SUPPORT

Need Help with this product?

Our specialists are here to help you find the best product or part available for your application. Call or Email us and we will make sure you get the right product or part for the job.

Call Us At1-800-323-4340
Live chat