Ricca Chemical Company HydroSpec™ Solvent K Aldehyde / Ketone Medium, one or two component volumetric titrations; 1 L

Representative image only.
Ricca Chemical Company Mfr # RK340000-1C – Item # EW-86998-43
Prepare any sample for analysis
  • Solvents and solvent blends dissolve even oily and fatty samples
  • Precisely formulated for stability and performance
Often bought with…

SPECS & DESCRIPTION

Specifications & Description

  • ClassReagent Solvents
  • FormLiquid
  • Volume (mL)1000
  • PackagingAmber Glass
  • DOT Information1-Methoxy-2-propanol solution; Class 3, Packing Group III
  • CAS NumberIncludes 107-98-2, 67-66-3, 7446-09-5
  • UN NumberUN3092
  • Shelf Life (days)1800
  • DescriptionSolvent K Aldehyde / Ketone Medium, one or two component volumetric titrations; 1 L
CorrosiveFlammableSolventToxic

Key Features

  • Solvents and solvent blends dissolve even oily and fatty samples
  • Precisely formulated for stability and performance

More About this Item

These Karl Fischer solvents and solvent blends offer sample preparation without trial and error. In addition to the standard methanol, there are blends featuring chloroform, xylene, 1-decanol, and propylene glycol monomethyl ether (PGME) for samples immiscible in pure methanol. These highly stable solvents retain activity for up to 5 years and bottles have the GL45 threaded neck to make connection to Karl Fischer titrators easy and simple.

REVIEWS

Q&A

CUSTOMER SUPPORT

Need Help with this product?

Our specialists are here to help you find the best product or part available for your application. Call or Email us and we will make sure you get the right product or part for the job.

Call Us At1-800-323-4340
Live chat