Ricca Chemical Company PAG10KN-100 Silver Plasma Grade Reference Standard, 1 mL = 10 mg Ag, 10,000 ppm Ag (120 mL)

Representative image only.
Ricca Chemical Company Mfr # PAG10KN-100 – Item # EW-86992-15
  • SILVER ICP STD., 1 = 10 Ag
  • Equivalent to 10,000 ppm

SPECS & DESCRIPTION

Specifications & Description

  • Purity GradePlasma
  • ClassSpectroscopy
  • ChemistryStandards
  • FormLiquid
  • Volume (mL)100
  • PackagingAmber Poly Bottle
  • DOT InformationCorrosive Liquid, Acidic, Inorganic, n.o.s., (Nitric Acid); Class 8, Packing Group III
  • CAS NumberIncludes 7697-37-2, 7732-18-5, and 7761-88-8
  • UN NumberUN3264
  • Shelf Life (months)18
  • DescriptionSilver Plasma Grade Reference Standard, 1 mL = 10 mg Ag, 10,000 ppm Ag (120 mL)

Key Features

  • SILVER ICP STD., 1 = 10 Ag
  • Equivalent to 10,000 ppm

More About this Item


Chemicals sold by Cole-Parmer are not intended for and should not be (i) used for medical, surgical or other patient oriented applications, or (ii) combined with any food product or ingested in any form.

REVIEWS

Q&A

CUSTOMER SUPPORT

Need Help with this product?

Our specialists are here to help you find the best product or part available for your application. Call or Email us and we will make sure you get the right product or part for the job.

Call Us At1-800-323-4340
Live chat