Ricca Chemical Company PSE10KN-100 Selenium Plasma Grade Reference Standard, 1 mL = 10 mg Se, 10,000 ppm Se, 120 mL

Representative image only.
Fisher Chemical Mfr # PSE10KN-100 – Item # EW-86992-04
Quality and consistency your lab can count on
  • Reagents and solutions to meet your lab and research needs
  • SELENIUM ICP STD., 1 = 10 Se
  • Equivalent to 10,000 ppm

SPECS & DESCRIPTION

Specifications & Description

  • Purity GradePlasma
  • ClassSpectroscopy
  • ChemistryStandards
  • FormLiquid
  • Volume (mL)100
  • PackagingPoly Bottle
  • DOT InformationCorrosive Liquid, Acidic, Inorganic, n.o.s., (Nitric Acid); Class 8, Packing Group III
  • CAS NumberIncludes 7697-32-2, 7732-18-5, and 7782-49-2
  • UN NumberUN3264
  • Shelf Life (months)18
  • DescriptionSelenium Plasma Grade Reference Standard, 1 mL = 10 mg Se, 10,000 ppm Se, 120 mL

Key Features

  • Reagents and solutions to meet your lab and research needs
  • SELENIUM ICP STD., 1 = 10 Se
  • Equivalent to 10,000 ppm

More About this Item

Chemicals sold by Cole-Parmer are not intended for and should not be (i) used for medical, surgical or other patient oriented applications, or (ii) combined with any food product or ingested in any form.

REVIEWS

Q&A

CUSTOMER SUPPORT

Need Help with this product?

Our specialists are here to help you find the best product or part available for your application. Call or Email us and we will make sure you get the right product or part for the job.

Call Us At1-800-323-4340
Live chat