Ricca Chemical Company PSI1KN-100 Silicon Plasma Grade Reference Standard, 1 mL = 1 mg Si, 1000 ppm (120 mL)

Representative image only.
Ricca Chemical Company Mfr # PSI1KN-100 – Item # EW-86992-07
  • SILICON ICP STD., 1 = 1 Si
  • Equivalent to 1,000 ppm

SPECS & DESCRIPTION

Specifications & Description

  • Purity GradePlasma
  • ClassSpectroscopy
  • ChemistryStandards
  • FormLiquid
  • Volume (mL)100
  • PackagingPoly Bottle
  • DOT InformationCorrosive Liquid, Acidic, Inorganic, n.o.s., (Nitric Acid and Hydrofluoric Acid); Class 8, Packing Group III
  • UN NumberUN3264
  • CAS NumberIncludes 7440-21-3, 7664-39-3, 7697-37-2, and 7732-18-5
  • Shelf Life (months)18
  • DescriptionSilicon Plasma Grade Reference Standard, 1 mL = 1 mg Si, 1000 ppm (120 mL)

Key Features

  • SILICON ICP STD., 1 = 1 Si
  • Equivalent to 1,000 ppm

More About this Item

Warning: Chemicals sold by Cole-Parmer are not approved or intended for, and should not be (i) used for medical, clinical, surgical or other patient oriented applications, or (ii) combined with any food product or ingested in any form.

REVIEWS

Q&A

CUSTOMER SUPPORT

Need Help with this product?

Our specialists are here to help you find the best product or part available for your application. Call or Email us and we will make sure you get the right product or part for the job.

Call Us At1-800-323-4340
Live chat