Ricca Chemical Company RV010043-500N Rhenium Standard for AAS 1000 ppm in 5% HNO₃; 500 mL

Representative image only.
Ricca Chemical Company Mfr # RV010043-500N – Item # EW-85001-17
Single-element calibration standard
  • Made to Guide 34 Standards!
  • Manufactured and Tested in an ISO 17025/Guide 34 Accredited Facility

SPECS & DESCRIPTION

Specifications & Description

  • Concentration1000 ppm
  • Purity GradeISO 17025 and ISO Guide 34
  • ClassSpectroscopy
  • ChemistryStandards
  • Medium5% Nitric Acid
  • Element/IonRhenium
  • Volume (mL)500
  • PackagingPoly Bottle
  • DOT InformationPacking Group 2, Packing Class 8, Corrosive Liquid Acidic Inorganic n.o.s. Nitric Acid
  • UN Number3264
  • Shelf Life (days)720
  • Shelf Life (months)24
  • DescriptionRhenium Standard for AAS 1000 ppm in 5% HNO₃; 500 mL

Key Features

  • Made to Guide 34 Standards!
  • Manufactured and Tested in an ISO 17025/Guide 34 Accredited Facility

More About this Item

Chemicals sold by Cole-Parmer® are not intended for and should not be (i) used for medical, surgical or other patient oriented applications, or (ii) combined with any food product or ingested in any form. Reagents utlilizes the highest quality raw materials to meet of exceed the specifications of the American Chemical Society.

For sale in the US only

REVIEWS

Q&A

CUSTOMER SUPPORT

Need Help with this product?

Our specialists are here to help you find the best product or part available for your application. Call or Email us and we will make sure you get the right product or part for the job.

Call Us At1-800-323-4340
Live chat