Ricca Chemical Company S5601000-500N Manganese AA Std, 1000 ppm Mn, Acetate in Nitric, 500 mL

Representative image only.
Ricca Chemical Company Mfr # S5601000-500N – Item # EW-86996-23
Quality and consistency your lab can count on
  • 1 ml = 0.05 mg Mn

SPECS & DESCRIPTION

Specifications & Description

  • ClassSpectroscopy
  • ChemistrySolvents
  • FormLiquid
  • Volume (mL)500
  • PackagingPoly Bottle
  • DOT InformationCorrosive Liquid, Acidic, Inorganic, n.o.s., (Nitric Acid); Class 8, Packing Group III
  • UN NumberUN3264
  • Shelf Life (months)24
  • DescriptionManganese AA Std, 1000 ppm Mn, Acetate in Nitric, 500 mL

Key Features

  • 1 ml = 0.05 mg Mn

More About this Item

Chemicals sold by Cole-Parmer are not intended for and should not be (i) used for medical, surgical or other patient oriented applications, or (ii) combined with any food product or ingested in any form.


Chemicals sold by Cole-Parmer are not intended for and should not be (i) used for medical, surgical or other patient oriented applications, or (ii) combined with any food product or ingested in any form.

REVIEWS

Q&A

CUSTOMER SUPPORT

Need Help with this product?

Our specialists are here to help you find the best product or part available for your application. Call or Email us and we will make sure you get the right product or part for the job.

Call Us At1-800-323-4340
Live chat