Ricca Chemical Company S8200000-500N Samarium AA Std, 1000 ppm Sm, Oxide in Nitric, 500 mL

Representative image only.
Ricca Chemical Company Mfr # S8200000-500N – Item # EW-86997-26

SPECS & DESCRIPTION

Specifications & Description

  • ClassSpectroscopy
  • ChemistryStandards
  • FormLiquid
  • Volume (mL)500
  • PackagingPoly Bottle
  • DOT InformationCorrosive Liquid, Acidic, Inorganic, n.o.s., (Nitric Acid); Class 8, Packing Group III
  • UN NumberUN3264
  • Shelf Life (months)24
  • DescriptionSamarium AA Std, 1000 ppm Sm, Oxide in Nitric, 500 mL

REVIEWS

Q&A

CUSTOMER SUPPORT

Need Help with this product?

Our specialists are here to help you find the best product or part available for your application. Call or Email us and we will make sure you get the right product or part for the job.

Call Us At1-800-323-4340
Live chat