Ricca Chemical Company S8245000-100N Scandium AA Std, 1000 ppm Sc, Oxide in Nitric, 100 mL

Representative image only.
Ricca Chemical Company Mfr # S8245000-100N – Item # EW-86997-29

SPECS & DESCRIPTION

Specifications & Description

  • ClassSpectroscopy
  • ChemistryStandards
  • FormLiquid
  • Volume (mL)100
  • PackagingPoly Bottle
  • DOT InformationCorrosive Liquid, Acidic, Inorganic, n.o.s., (Nitric Acid); Class 8, Packing Group III
  • UN NumberUN3264
  • Shelf Life (months)24
  • DescriptionScandium AA Std, 1000 ppm Sc, Oxide in Nitric, 100 mL

REVIEWS

Q&A

CUSTOMER SUPPORT

Need Help with this product?

Our specialists are here to help you find the best product or part available for your application. Call or Email us and we will make sure you get the right product or part for the job.

Call Us At1-800-323-4340
Live chat