Ricca Chemical Company VeriSpec™ CDRL Standard 22; 100 mL

Representative image only.
Ricca Chemical Company Mfr # RV010837-100N – Item # EW-85013-44
Multi-element/ion calibration solutions
  • Made to Guide 34 Standards!
  • Manufactured and Tested in an ISO 17025/Guide 34 Accredited Facility
  • 500 ppm: Ca, K, Mg, Na; 20 ppm Al, Ba; 10 ppm Fe, 6 ppm Sb; 5 ppm Co, V; 4 ppm Ni, 2.5 ppm Cu, 2 ppm Zn, 1.5 ppm Mn; 1 ppm Ag, As, Cr, Tl; 0.5 ppm Be, Cd, Se; 0.3 ppm Pb

SPECS & DESCRIPTION

Specifications & Description

  • ConcentrationAg 1 ppm, Al 20 ppm, As 1 ppm, Ba 20 ppm, Be 0.5 ppm, Ca 500 ppm, Cd 0.5 ppm, Co 5 ppm, Cr 1 ppm, Cu 2.5 ppm, Fe 10 ppm, K 500 ppm, Mg 500 ppm, Mn 1.5 ppm, Na 500 ppm, Ni 4 ppm, Pb 0.3 ppm, Sb 6 ppm, Se 0.5 ppm, Tl 1 ppm, V 5 ppm, and Zn 2 ppm
  • Purity GradeISO 17025 and ISO Guide 34
  • ClassSpectroscopy
  • Medium5% Nitric Acid
  • ChemistryStandards
  • Element/IonSilver, Aluminum, Arsenic, Barium, Beryllium, Calcium, Cadmium, Cobalt, Chromium, Copper, Iron, Potassium, Magnesium, Manganese, Sodium, Nickel, Lead, Antimony, Selenium, Thallium, Vanadium, and Zinc
  • Volume (mL)100
  • PackagingPoly Bottle
  • DOT InformationPacking Group 2, Packing Class 8, Corrosive Liquid Acidic Inorganic n.o.s. Nitric Acid
  • UN Number3264
  • Shelf Life (days)180
  • Shelf Life (months)6
  • DescriptionCDRL Standard 22; 100 mL

Key Features

  • Made to Guide 34 Standards!
  • Manufactured and Tested in an ISO 17025/Guide 34 Accredited Facility
  • 500 ppm: Ca, K, Mg, Na; 20 ppm Al, Ba; 10 ppm Fe, 6 ppm Sb; 5 ppm Co, V; 4 ppm Ni, 2.5 ppm Cu, 2 ppm Zn, 1.5 ppm Mn; 1 ppm Ag, As, Cr, Tl; 0.5 ppm Be, Cd, Se; 0.3 ppm Pb

More About this Item

For sale in the US only

REVIEWS

Q&A

CUSTOMER SUPPORT

Need Help with this product?

Our specialists are here to help you find the best product or part available for your application. Call or Email us and we will make sure you get the right product or part for the job.

Call Us At1-800-323-4340
Live chat