Ricca Chemical Company VeriSpec™ Mixed Anion Standard 5; 100 mL

Representative image only.
Ricca Chemical Company Mfr # RV010688-100N – Item # EW-85011-95
Multi-element/ion calibration solutions
  • Made to Guide 34 Standards!
  • Manufactured and Tested in an ISO 17025/Guide 34 Accredited Facility
  • 150 ppm SO₄²-, PO₄³; 100 ppm NO₃-, 30 ppm Cl-, 20 ppm F-

PRODUCT OPTIONS

$217.00USD / Each

SPECS & DESCRIPTION

Specifications & Description

  • ConcentrationCl⁻ 30 ppm, F⁻ 20 ppm, NO₃⁻ 100 ppm, PO₄³⁻ 150 ppm, and SO₄²⁻ 150 ppm
  • Purity GradeISO 17025 and ISO Guide 34
  • ClassSpectroscopy
  • ChemistryStandards
  • MediumWater
  • Element/IonChloride, Fluoride, Nitrate, Phosphate, and Sulfate
  • Volume (mL)100
  • PackagingPoly Bottle
  • DOT InformationNot regulated by DOT
  • Shelf Life (days)180
  • Shelf Life (months)6
  • DescriptionMixed Anion Standard 5; 100 mL

Key Features

  • Made to Guide 34 Standards!
  • Manufactured and Tested in an ISO 17025/Guide 34 Accredited Facility
  • 150 ppm SO₄²-, PO₄³; 100 ppm NO₃-, 30 ppm Cl-, 20 ppm F-

More About this Item

For sale in the US only

ACCESSORIES

REVIEWS

Q&A

CUSTOMER SUPPORT

Need Help with this product?

Our specialists are here to help you find the best product or part available for your application. Call or Email us and we will make sure you get the right product or part for the job.

Call Us At1-800-323-4340
Live chat