Ricca Chemical Company VeriSpec™ Nexion KED ICP-MS Calibration Standard for Set Up 2; 500 mL

Representative image only.
Ricca Chemical Company Mfr # RV010882-500N – Item # EW-85013-87
Multi-element instrument calibration standards
  • Made to Guide 34 Standards!
  • Manufactured and Tested in an ISO 17025/Guide 34 Accredited Facility
  • 10 ppb Co, 1 pb Ce

SPECS & DESCRIPTION

Specifications & Description

  • Purity GradeISO 17025 and ISO Guide 34
  • ConcentrationCe 1 ppb and Co 10 ppb
  • ClassSpectroscopy
  • ChemistryStandards
  • Medium1% Hydrochloric Acid
  • Element/IonCerium and Cobalt
  • Volume (mL)500
  • PackagingPoly Bottle
  • DOT InformationNot regulated by DOT
  • Shelf Life (days)180
  • Shelf Life (months)6
  • DescriptionNexion KED ICP Calibration Standard for Set Up 2; 500 mL

Key Features

  • Made to Guide 34 Standards!
  • Manufactured and Tested in an ISO 17025/Guide 34 Accredited Facility
  • 10 ppb Co, 1 pb Ce

More About this Item

Chemicals sold by Cole-Parmer® are not intended for and should not be (i) used for medical, surgical or other patient oriented applications, or (ii) combined with any food product or ingested in any form. Reagents utlilizes the highest quality raw materials to meet of exceed the specifications of the American Chemical Society.

REVIEWS

Q&A

CUSTOMER SUPPORT

Need Help with this product?

Our specialists are here to help you find the best product or part available for your application. Call or Email us and we will make sure you get the right product or part for the job.

Call Us At1-800-323-4340
Live chat