Ricca Chemical Company VeriSpec™ Potassium Standard for AAS 1000 ppm in 2% HNO₃; 500 mL

Representative image only.
Ricca Chemical Company Mfr # RV010075-500N – Item # EW-85001-87
Single-element calibration standard
  • Made to Guide 34 Standards!
  • Manufactured and Tested in an ISO 17025/Guide 34 Accredited Facility

PRODUCT OPTIONS

$64.00USD / Each
Often bought with…

SPECS & DESCRIPTION

Specifications & Description

  • Concentration1000 ppm
  • Purity GradeISO 17025 and ISO Guide 34
  • ClassSpectroscopy
  • ChemistryStandards
  • Medium2% Nitric Acid
  • Element/IonPotassium
  • Volume (mL)500
  • PackagingPoly Bottle
  • DOT InformationNot regulated by DOT
  • Shelf Life (days)720
  • Shelf Life (months)24
  • DescriptionPotassium Standard for AAS 1000 ppm in 2% HNO₃; 500 mL

Key Features

  • Made to Guide 34 Standards!
  • Manufactured and Tested in an ISO 17025/Guide 34 Accredited Facility

More About this Item

For sale in the US only

ACCESSORIES

REVIEWS

Q&A

CUSTOMER SUPPORT

Need Help with this product?

Our specialists are here to help you find the best product or part available for your application. Call or Email us and we will make sure you get the right product or part for the job.

Call Us At1-800-323-4340
Live chat