Ricca Chemical Company VeriSpec™ Ruthenium Standard for ICP 1000 ppm in 5% HCl; 50 mL

Representative image only.
Ricca Chemical Company Mfr # RV010308-50C – Item # EW-85004-74
Single-element calibration standard
  • Made to Guide 34 Standards!
  • Manufactured and Tested in an ISO 17025/Guide 34 Accredited Facility

SPECS & DESCRIPTION

Specifications & Description

  • Concentration1000 ppm
  • Purity GradeISO 17025 and ISO Guide 34
  • ClassSpectroscopy
  • ChemistryStandards
  • Medium5% Hydrochloric Acid
  • Element/IonRuthenium
  • Volume (mL)50
  • PackagingPoly Bottle
  • DOT InformationPacking Group 2, Packing Class 8, Corrosive liquid acidic inorganic n.o.s. Hydrochloric Acid Ruthenium Trichloride
  • UN Number3264
  • Shelf Life (days)720
  • Shelf Life (months)24
  • DescriptionRuthenium Standard for ICP 1000 ppm in 5% HCl; 50 mL

Key Features

  • Made to Guide 34 Standards!
  • Manufactured and Tested in an ISO 17025/Guide 34 Accredited Facility

More About this Item

For sale in the US only

REVIEWS

Q&A

CUSTOMER SUPPORT

Need Help with this product?

Our specialists are here to help you find the best product or part available for your application. Call or Email us and we will make sure you get the right product or part for the job.

Call Us At1-800-323-4340
Live chat