Richard-Allan Scientific Cytoseal Cytoseal 60 Histology Mounting Media, 4 oz Bottles; 12/Cs
Representative image only.
Richard-Allan Scientific Cytoseal Mfr # 8310-4 – Item # EW-48802-01
  • Not for sale outside of the United States
PRODUCT OPTIONS
$296.75USD / Case of 12
SPECS & DESCRIPTION
Specifications & Description
  • Capacity (mL)118
  • DescriptionCytoseal 60 Histology Mounting Media, 4 oz Bottles; 12/Cs
Key Features
  • Not for sale outside of the United States
More About this Item
Clear, colorless media contains an anti-oxidant to prevent yellowing or fading of stained sections. Media come in an easy-to-use dispenser bottle — prevent spills while promoting precise dispensing. Feature the same refractive index as immersion oil. Sold in case of 12.