Dynalon Round Dipping Basket, PP for Crock 06724-20
Representative image only.
Dynalon Mfr # 167174-20 – Item # EW-06717-20
  • 1 year warranty
PRODUCT OPTIONS
$336.25USD / Each
BUY MORE & SAVE
  • 1-5$336.25USD / Each
  • 6-29$302.63USD / Each
  • 30-119$285.81USD / Each
  • 120+$269.00USD / Each
SPECS & DESCRIPTION
Specifications & Description
  • MaterialPolypropylene
  • Overall Height (in)8
  • Overall Height (cm)20.3
  • Inside Diameter (ID) (in)8
  • Inside Diameter (ID) (cm)20.3
  • DescriptionRound Dipping Basket, PP for Crock 06724-20
  • Warranty1 year
1 Year WarrantyNSF international certified
Key Features
  • 1 year warranty
More About this Item
These polypropylene (PP) baskets withstand temperatures to 250°>F (121°>C). Sides and bottom have 1/8'' perforations. Reinforced top and bottom rings give added strength. Swing-type handle. Use with 06724-series Heavy Crocks.