Rubberfab Gold-colored IDentifier Hinge Clamp, 1/2-3/4" Clamp Size, 304 SS
Representative image only.
Rubberfab Mfr # 13MHHM-304-050/075-GOLD – Item # EW-30562-51
Easy visual identification of critical joints for review in your process
  • Contructed of gold-colored 304 Stainless Steel
PRODUCT OPTIONS
$48.50USD / Each
SPECS & DESCRIPTION
Specifications & Description
  • Fitting MaterialTitanium Nitride
  • Connection TypeSanitary
  • Sanitary Clamp Size (Port 1) (in)3/4
  • DescriptionGold-colored IDentifier Hinge Clamp, 1/2-3/4" Clamp Size, 304 SS
Key Features
  • Contructed of gold-colored 304 Stainless Steel
More About this Item
Easily distinguishable from standard stainless steel clamps from a distance, these gold-colored sanitary clamps are identical in size and function and can be used interchangeably with standard stainless steel clamps. Clamps feature a TiN (titanium nitride) coating. Clamps are available in a variety of sizes.