Rubberfab TR-50-TEE-CP Torque Tee Sanitary Clamp Securing Handle, 50 in-lb
Representative image only.
Rubberfab Mfr # TR-50-TEE-CP – Item # EW-23508-03
Ensure the security of sanitary connections with preset torque output
  • Works with any sanitary clamp wing nut and Rubberfab's Torque-Rite® nut
  • Easily reach into tight or awkward spaces
  • Choose the appropriate torque output per the clamp manufacturer's specifications
  • Audible click alerts you when the torque has been reached
  • Ratcheting mechanism disengages for easy clamp removal and is sealed within handle for durability
PRODUCT OPTIONS
$203.00USD / Each
SPECS & DESCRIPTION
Specifications & Description
  • Drive (in)0.25
  • Drive (in-lb)50
  • DescriptionTorque Tee Sanitary Clamp Securing Handle, 50 in-lb
Key Features
  • Works with any sanitary clamp wing nut and Rubberfab's Torque-Rite® nut
  • Easily reach into tight or awkward spaces
  • Choose the appropriate torque output per the clamp manufacturer's specifications
  • Audible click alerts you when the torque has been reached
  • Ratcheting mechanism disengages for easy clamp removal and is sealed within handle for durability
  • "T" shape creates effective leverage for fast, easy one-handed tightening
  • Recommended for PTFE, Tuf-Steel®, and Envelope Gaskets
More About this Item
The Rubberfab Torque Tee and universal socket are designed to safely and securely install and remove sanitary clamps. Available with a family of components, the Torque Tee handle creates effective tightening leverage, even in hard-to-reach spaces. If you need to loosen or remove the clamp, simply disengage the ratcheting mechanism to lock the handle. The universal socket is required when using the Torque Tee.
Handles require the universal socket (23508-08) to function with standard sanitary clamp wing nuts.