Saint Gobain Graduated Container/Tank with Cover, HDPE, 50 gallon
Representative image only.
Saint Gobain Mfr # 56104-0050 – Item # EW-06972-18
Tough and corrosion-resistant
  • Rigid yet lightweight
  • Straight, no-lip flange is easy to clean
  • Fit through standard doors for easy portability
  • 1 year warranty
SPECS & DESCRIPTION
Specifications & Description
  • MaterialHDPE
  • Volume (Gal)50
  • Volume (Liters)189
  • Height (in)22 1/2
  • Height (cm)57.15
  • Outside Diameter (OD) (in)22
  • Graduations (Gal)5
  • Graduations (Liters)17
  • DescriptionGraduated Container/Tank with Cover, HDPE, 50 gallon
  • Warranty1 Year
1 Year Warranty
Key Features
  • Rigid yet lightweight
  • Straight, no-lip flange is easy to clean
  • Fit through standard doors for easy portability
  • 1 year warranty
More About this Item
These multi-purpose HDPE tanks are seamlessly designed with high-quality virgin resin and eliminate the major causes of leakage associated with lesser quality tanks. Tanks wont wick or crack, and have a visible liquid level graduations. Tanks feature rounded corners that have less molded-in stress, which prolongs life of the tank.