Salisbury by Honeywell GK0011B8 Insulated Glove Kit, Class 00, Black, 11"L, Size 8; One Pair
Representative image only.
Salisbury by Honeywell Mfr # GK0011B8 – Item # EW-86435-05
Comply with NFPA 70E standard while protecting employees from dangerous electrical injuries
PRODUCT OPTIONS
$191.00USD / Each
SPECS & DESCRIPTION
Specifications & Description
  • MaterialRubber
  • Size8
  • Length (in)11
  • Length (cm)27.9
  • ColorBlack
  • ComplianceClass 00
  • SterileNo
  • DescriptionInsulated Glove Kit, Class 00, Black, 11"L, Size 8; One Pair
Includes
insulating rubber glove, leather protector glove, and storage bag.
More About this Item
Get everything you need to protect yourself and keep your insulating rubber gloves in top working condition. Kits include a pair of insulating rubber gloves; the appropriate leather protector that protects the glove from cuts, abrasions, and punctures; and a storage bag which keeps the glove flat, preventing folds and creases that strain the rubber, shortening the life of the glove.
Note: In accordance with NFPA 70E, anything over 50 volts requires insulating rubber gloves.