SalvisLab 31F04210 Wire Shelf for Coolingcenter CIC150 Refrigerated Incubator
Representative image only.
SalvisLab Mfr # 31F04210 – Item # EW-39701-55
Precise temperature control for routine lab test or long-term operation
  • Precise control of fan speed from 60 to 100%
  • Storage of up to 50 programs and 15 program steps
  • Temperature regulated by quality PT 100 temperature sensors
  • 2 year warranty
SPECS & DESCRIPTION
Specifications & Description
  • Temperature Accuracy±0.2⁰C at 37⁰C
  • DescriptionWire Shelf for Coolingcenter CIC150 Refrigerated Incubator
  • Warranty2 year
Two year warranty
Key Features
  • Precise control of fan speed from 60 to 100%
  • Storage of up to 50 programs and 15 program steps
  • Temperature regulated by quality PT 100 temperature sensors
  • 2 year warranty
More About this Item
SalvisLab Coolingcenter is the perfect refrigerated incubator for every routine test in the lab including high batch operations over long periods. Exclusive EasyMenu microprocessor controller features a digital LCD with easy-to-operate keypad, menu prompts in five selectable languages, intelligent fan speed control, real-time clock, timer, and RS-232 output.
Rugged steel exterior combined with robust insulation minimizes heat/cooling loss, saving energy and keeping exterior temperature at ambient. Mechanical switch on door prevents unit from running when door is open; as soon as it is closed, process will start running again. Over temperature protection shut down operation if max temperature is exceeded.